Learn More
Apexbio Technology LLC THZ1 Hydrochloride, 25mg. Cas: 1604810-83-4 (free base) MFCD: MFCD28167786. CDK7 inhibitor
SDP
Supplier:  Apexbio Technology LLC B473625
Encompass_Preferred
THZ1 is a covalent inhibitor of CDK7 with IC50 value of 3.2nM [1].THZ1 covalently modifies CDK7 by targeting C312 residue outside of the kinase domain, providing an unanticipated means of achieving covalent selectivity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5356