Learn More
Selleck Chemical LLC THZ2
SDP
Supplier:  Selleck Chemical LLC S79692MG
Encompass_Preferred
THZ2 a THZ1 analog is a selective inhibitor of CDK7 with IC50 of 13 9 nM THZ2 efficiently suppresses the clonogenic growth of TNBC cells with IC50 of 10 nM
Catalog No. 50-000-02833