Learn More
Selleck Chemical LLC THZ531 5mg 1702809-17-3
SDP
Supplier:  Selleck Chemical LLC S65955MG
Encompass_Preferred
THZ531 is a selective covalent inhibitor of CDK12 and CDK13 with IC50 values of 158 and 69 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1826507