Learn More
Medchemexpress LLC Tiagabine hydrochloride | 145821-59-6 | MFCD07369025 | 99.4% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0696A50MG
Encompass_Preferred
Tiagabine hydrochloride is an anticonvulsant agent and a selective inhibitor of the GAT-1 GABA transporter, with a Ki of approximately 0.1 μM. It potently and selectively inhibits GABA reuptake with IC50s of 67, 446, and 182 nM for [3H]GABA uptake in synaptosomes, neurons, and glia. This compound also exhibits neuroprotective properties.
  • Selective inhibitor of GAT-1 GABA transporter
  • Potently and selectively inhibits GABA reuptake
  • Exhibits neuroprotective properties
  • Protects neurons against mutant huntingtin-induced toxicity
  • Increases viability of SH-SY5Y cells
  • Neuroprotective in mouse models of Huntington's disease
  • Reduces dopaminergic neuronal loss in PD mouse models
  • Shows anti-allodynic effect in a rat model of neuropathic pain
  • Antinociceptive effects against acute thermal pain in mice
  • Suppresses pentylenetetrazole-induced seizures and improves cognitive parameters in mice
Catalog No. 50-004-05502