Learn More
TARGETMOL CHEMICALS INC Tiamulin fumarate 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T353461G
Encompass_Preferred
Also available in 1 mL, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tiamulin fumarate (Thiamutilin fumarate) is a semisynthetic diterpenoid antibiotic with antibacterial activity and inhibition of protein synthesis, which can be used to study pneumocystis. Purity 99.64%
Catalog No. 50-287-6452