Learn More
Apexbio Technology LLC Tiamulin fumarate 55297-96-6 250mg
SDP
Supplier:  Apexbio Technology LLC A9910250
Encompass_Preferred
Tiamulin fumarate is a semi-synthetic antibiotic belonging to the pleuromutilin class specifically inhibiting bacterial protein synthesis Its primary antibacterial activity targets Gram-positive bacteria certain Gram-negative bacteria and mycoplasmas by binding to the bacterial 50S ribosomal subunit thereby blocking peptide chain elongation Laboratory studies commonly utilize Tiamulin fumarate for investigating bacterial susceptibility mechanisms antibiotic resistance profiles and antimicrobial efficacy in veterinary pathogens It is applied predominantly in veterinary research settings involving swine mycoplasmal pneumonia swine dysentery and chronic respiratory infections in poultry In vitro assays demonstrate antibacterial activity with reported minimum inhibitory concentration (MIC) values often in the low micromolar range and inhibitory concentrations (IC50) varying depending on bacterial strain and experimental conditions
Catalog No. 50-000-62321