Learn More
Cayman Chemical TIamulInfumarate 5g
SDP
Supplier:  Cayman Chemical 212165 G
Encompass_Preferred
A pleuromutilin antibiotic; active against various mycobacteria (MICs = 0.0025-0.25 UG/ml) and various Gram-positive bacteria, including MRSA; binds to the peptidyl transferase in the 50S ribosomal subunit to inhibit protein synthesis
Catalog No. 50-298-1029