Learn More
TARGETMOL CHEMICALS INC Tiapamil hydrochloride 1MG
Supplier: TARGETMOL CHEMICALS INC T131541MG
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Tiapamil hydrochloride (Ro 11-1781 hydrochloride) is a calcium channel blocker with antihypertensive activity and may be used in the study of angina and cardiovascular disease. Purity 98.47%