Learn More
Medchemexpress LLC Tiapride hydrochloride | 51012-33-0 | MFCD00133861 | 99.95% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB119610MM/1ML
Encompass_Preferred
Tiapride hydrochloride is a selective and orally active antagonist of D2 and D3 dopamine receptors, with IC50 values ranging from 110-320 nM for D2 and 180 nM for D3 receptors. It exhibits anti-dyskinetic and anxiolytic activities and is classified as a neuroleptic agent.
  • Selective D2 and D3 dopamine receptor antagonist.
  • Orally active.
  • Shows anti-dyskinetic activity.
  • Exhibits anxiolytic activity.
  • Neuroleptic agent.
  • Antagonizes ethanol-induced hyperactivity in mouse models without affecting sleep time.
Catalog No. 50-004-12421