Learn More
Cayman Chemical TIaprofenIc AcId 10mg
SDP
Supplier:  Cayman Chemical 2736410 MG
Encompass_Preferred
A COX inhibitor and NSAID with anti-inflammatory and analgesic activities; inhibits synthesis of PGE2 and PGF2α from arachidonic acid in bovine seminal vesicle microsomes and inhibits TXB2 formation in human umbilical cord arteries bathed in clotting human blood (IC50 = 0.71 μM); has anti-inflammatory activity in rat models of carrageenan-induced edema and adjuvant-induced arthritis and has analgesic activity in acetic acid- or phenylquinone-induced writhing tests in rodents
Catalog No. 50-294-0158