Learn More
Medchemexpress LLC Tiaprofenic acid | 33005-95-7 | MFCD00866089 | 99.9% | 260.31 g/mol | C14H12O3S | 1 ML

Supplier: Medchemexpress LLC HY10657910MM 1ML
Tiaprofenic acid (CAS 33005-95-7) is a nonsteroidal anti-inflammatory compound supplied for research use. It is available as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes. The material is high purity and intended for in vitro pharmacology, assay development, and analytical work.
- Ready-to-use 10 mM solution in DMSO, 1 mL volume.
- High purity (~99.9%) suitable for research applications.
- Molecular formula C14H12O3S, molecular weight 260.31 g/mol.
- Available in solid forms for formulation flexibility.
- Intended for research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.