Learn More
Enzo Life Sciences 2-Thio(3-iodobenzyl)-5-(1-pyridyl)-[1,3,4]-oxadiazole (5mg). CAS: 478482-75-6
SDP
Supplier:  Enzo Life Sciences ALX270358M005
Encompass_Preferred
Potent inhibitor of glycogen synthase kinase-3β (GSK-3β) (IC50=390nM). Alternative name: TIBPO. Formula: C14H10IN3OS. MW: 395.2. Formulation: Lyophilized. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8505