Learn More
Medchemexpress LLC Ticagrelor impurity 10 | 2803420-77-9 | 98.5% | C26H32F2N6O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY77732A5MG
Encompass_Preferred
Ticagrelor impurity 10 is an impurity of Ticagrelor, intended for research use only. This chemical reagent has a molecular weight of 562.63 and appears as a white to off-white solid.
  • Impurity of ticagrelor
  • For research use only
  • Molecular weight of 562.63
  • Appears as a white to off-white solid
  • Shipped at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-34944