Learn More
Medchemexpress LLC Ticagrelor impurity 11 | 220352-36-3 | ≥95.0% | C10H8F2O2 | 100 MG

Supplier: Medchemexpress LLC HY77964100MG
Ticagrelor impurity 11 is an impurity of Ticagrelor. It has a molecular weight of 198.17 and appears as a solid. This product is for research use only.
- Purity of ≥95.0%
- Chemical formula: C10H8F2O2
- Molecular weight: 198.17
- Appears as a solid
- Suitable for research use only
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.