Learn More
Medchemexpress LLC Ticagrelor impurity 11 | 220352-36-3 | ≥95.0% | C10H8F2O2 | 250 MG

Supplier: Medchemexpress LLC 250MGHY77964
MedChemExpress HY-77964, also known as Ticagrelor impurity 11, is an impurity of Ticagrelor (HY-10064). It is a solid compound with a molecular weight of 198.17 and a purity of ≥95.0%. This chemical is intended for laboratory use and the manufacture of substances.
- Appearance: Solid
- Chemical formula: C10H8F2O2
- Molecular weight: 198.17
- CAS number: 220352-36-3
- Purity (NMR): ≥95.0%
- Storage for powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- Shipping: Room temperature if less than 2 weeks
- Identified uses: Laboratory chemicals, manufacture of substances
- Related classifications: Drug intermediate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.