Learn More
Medchemexpress LLC Ticagrelor impurity 12 | 220241-61-2 | 99.6% | 385.88 | C15H20ClN5O3S | 10 MG

Supplier: Medchemexpress LLC HY7854010MG
Ticagrelor impurity 12 is an analytical standard used as an impurity reference for ticagrelor in research and analytical testing. The compound is characterized by high purity and defined physicochemical properties for method development and quality control.
- High purity standard (99.6%) for analytical assays.
- Molecular weight 385.88 g/mol.
- CAS number 220241-61-2 for unambiguous identification.
- Recommended storage under nitrogen at -20°C; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
- Intended for research and analytical use (not for human consumption).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.