Learn More
Medchemexpress LLC Ticagrelor impurity 14 | 2408330-73-2 | MFCD25977366 | 98.0% | 434.94 g/mol | C17H27ClN4O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY7814810MG
Encompass_Preferred
Ticagrelor impurity 14 is a defined small-molecule impurity of the antiplatelet agent ticagrelor, supplied for laboratory research and analytical use. It is provided in small pack sizes suitable for impurity profiling, method development, and related analytical studies. CAS: 2408330-73-2; molecular formula: C17H27ClN4O5S; molecular weight: 434.94 g/mol. Recommended storage: -20°C sealed; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Suitable for impurity profiling and analytical method development.
  • Provided in small quantities for laboratory research and validation.
  • Characterized by CAS 2408330-73-2 and molecular formula C17H27ClN4O5S.
  • Recommended storage: -20°C sealed; in solvent, -80°C for 6 months or -20°C for 1 month.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-98695