Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 16 | 2096989-56-7 | 99.5% | C23H28F2N6O4S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY784955MG

Encompass_Preferred

Ticagrelor impurity 16 is an impurity of Ticagrelor, intended for research use only. It is a solid with a molecular weight of 522.57.

  • Impurity of ticagrelor
  • Solid form
  • Molecular weight of 522.57
  • Intended for research use only
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years
  • Store in solvent at -20°C for 1 year

Catalog No. 50-004-33231


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.