Learn More
Medchemexpress LLC Ticagrelor impurity 26 | 1644461-85-7 | 99.4% | 538.57 g/mol | C23H28F2N6O5S | 10 MG

Supplier: Medchemexpress LLC HY7804610MG
Ticagrelor impurity 26 is a characterized impurity of the antiplatelet agent ticagrelor provided as a high-purity analytical reference material for impurity profiling and method development.
- Reported purity 99.39%.
- CAS number 1644461-85-7.
- Molecular formula C23H28F2N6O5S; molecular weight 538.57 g/mol.
- Intended for HPLC and LC-MS method development and impurity identification.
- Supplied in small quantities suitable for analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.