Learn More
Medchemexpress LLC Ticagrelor impurity 26 | 1644461-85-7 | 99.4% | C23H28F2N6O5S | 1 MG

Supplier: Medchemexpress LLC HY780461MG
Ticagrelor impurity 26 is a chemical compound used for research applications. It is identified by CAS number 1644461-85-7 and has a molecular formula of C23H28F2N6O5S. This compound is supplied as an off-white to light yellow solid and has a purity of 99.4% (LCMS).
- Purity (LCMS): 99.4%
- Molecular weight: 538.57
- Appearance: Off-white to light yellow solid
- Melting point: 206-208 °C
- Recommended powder storage: -20°C for 3 years or 4°C for 2 years
- Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.