Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 26 | 1644461-85-7 | 99.4% | C23H28F2N6O5S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY780465MG

Encompass_Preferred

Ticagrelor impurity 26 is an off-white to light yellow solid with a purity of 99.4%. This chemical is for research use only and intended for experienced personnel in appropriately equipped and authorized facilities. It is stable under recommended storage conditions, but incompatible with strong acids/alkalis, and strong oxidizing/reducing agents. Hazardous decomposition products may be emitted under fire conditions.

  • Off-white to light yellow solid appearance
  • Purity (LCMS) of 99.4%
  • Stable under recommended storage conditions
  • For research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-33210


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.