Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ticagrelor impurity 5 | 1863036-49-0 | >99.5% | 578.63 g/mol | C26H32F2N6O5S | 10 MG
SDP

Supplier:  Medchemexpress LLC HY7854610MG

Encompass_Preferred

Ticagrelor impurity 5 is an analytical standard used as an impurity reference for ticagrelor. It is provided for research and analytical applications including impurity profiling, method development, and quality control. The compound has molecular formula C26H32F2N6O5S and a molecular weight of 578.63 g/mol.

  • Analytical standard for impurity profiling and method development
  • High reported purity (>99.5%) suitable for HPLC and MS reference
  • Molecular formula C26H32F2N6O5S; molecular weight 578.63 g/mol
  • Suitable for identity confirmation and chromatographic reference materials
  • Recommended storage: solid at 4°C; in solvent under nitrogen at -80°C up to 6 months

Catalog No. 50-002-99147


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.