Learn More
Medchemexpress LLC Ticagrelor impurity 5 | 1863036-49-0 | C26H32F2N6O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY785465MG
Encompass_Preferred
Ticagrelor impurity 5 is an impurity of Ticagrelor (HY-10064). This product requires specific storage conditions to maintain stability and efficacy.
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for up to 6 months
  • When in solvent, store at -20°C for up to 1 month under nitrogen
Catalog No. 50-004-33235