Learn More
Medchemexpress LLC Ticagrelor impurity 6 | 2216753-87-4 | 99.1% | C23H28F2N6O4S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY780481MG
Encompass_Preferred
Ticagrelor impurity 6 is a high-purity chemical compound, presenting as a white to off-white solid. It has a molecular weight of 522.57 and a chemical formula of C23H28F2N6O4S. This substance serves as an impurity of Ticagrelor.
  • Impurity of ticagrelor
  • High purity (99.12%) for research applications
  • White to off-white solid appearance
  • Suitable for laboratory chemical uses
  • Identified as a drug intermediate
Catalog No. 50-004-34824