Learn More
Gold Biotechnology Inc Timentin™ Ticarcillin/Clavulanate (15/1) 10 g
Supplier: Gold Biotechnology Inc T10410
Timentin™ is a 15:1 mixture of ticarcillin and clavulanate. Ticarcillin is a penicillin ß-lactam antibiotic, which is susceptible to ß-lactamase degradation. When combined with clavulanate, a ß-lactamase inhibitor, its efficacy is greatly increased. Timentin™ exhibits high activity against gram-negative bacteria and resistant Agrobacterium species.