Learn More
Cayman Chemical TIclopIdIn-d4hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 334675 MG
Encompass_Preferred
An internal standard for the quantification of ticlopidine by GC- or LC-MS
Catalog No. 50-294-8461