Learn More
Apexbio Technology LLC Tideglusib, 10mg. Cas: 865854-05-3 MFCD: MFCD18633296. non-ATP-competitive GSK-3β inhibitor
SDP
Supplier:  Apexbio Technology LLC B153910
Encompass_Preferred
IC50: A potent, selective and irreversible non-ATP-competitive GSK-3 suppressor with an IC50 of 60 nM.Tideglusib is a GSK-3 inhibitor currently undergoing phase II clinical trials for Alzheimer disease and progressive supranuclear palsy. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5254