Learn More
Medchemexpress LLC Tie2 kinase inhibitor 1 | 948557-43-5 | MFCD18384977 | 439.53 g/mol | C26H21N3O2S | 50 MG

Supplier: Medchemexpress LLC HY10055650MG
Tie2 kinase inhibitor 1 is a small-molecule, ATP-site-targeting inhibitor selective for the Tie2 (TEK) receptor tyrosine kinase. It exhibits in vitro potency with reported anti-cancer activity and is supplied for research use only.
- Selective Tie2 kinase inhibition (IC50 250 nM).
- Small-molecule, cell-permeable inhibitor.
- Demonstrated anti-cancer activity in published studies.
- Supplied for research use only, not for human or clinical use.
- Molecular formula C26H21N3O2S; molecular weight 439.53 g/mol.
- CAS number 948557-43-5.
- Provided with SDS for safe handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.