Learn More
Medchemexpress LLC Tigecycline | 220620-09-7 | >98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB011750MG
Encompass_Preferred
Tigecycline (GAR-936) is a broad-spectrum glycylcycline antibiotic used as a laboratory chemical for the manufacture of substances. It exhibits a mean inhibitory concentration (MIC) of approximately 125 ng/mL for E. coli (MG1655 strain) and MIC50 and MIC90 values of 1 and 2 mg/L, respectively, for Acinetobacter baumannii. This product is intended for research use only and has not been fully validated for medical applications.
  • Broad-spectrum glycylcycline antibiotic
  • Effective against E. coli (MG1655 strain) with an MIC of approximately 125 ng/mL
  • Active against Acinetobacter baumannii with MIC50 and MIC90 values of 1 and 2 mg/L
  • Classified as acutely toxic if swallowed (Category 4)
  • Causes skin irritation (Category 2) and serious eye irritation (Category 2A)
  • May cause respiratory irritation (Category 3)
  • For research use only
Catalog No. 50-004-50751