Learn More
Medchemexpress LLC Tigecycline | 220620-09-7 | 99.95% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB011710MM/1ML
Encompass_Preferred
Tigecycline is a broad-spectrum glycylcycline antibiotic. It demonstrates inhibitory activity against various bacterial strains such as *E. coli* and *Acinetobacter baumannii*, and also inhibits AML2 and HL-60 cells in vitro. In vivo studies have shown that Tigecycline can reduce tumor volume and weight in acute myeloid leukemia xenograft models.
  • Broad-spectrum glycylcycline antibiotic
  • Inhibits *E. coli* and *A. baumannii*
  • Demonstrates in vitro activity against AML2 and HL-60 cells
  • Reduces tumor volume and weight in AML xenograft models
Catalog No. 50-004-50843