Learn More
Medchemexpress LLC Tigecycline | 220620-09-7 | 99.95% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB01175MG
Encompass_Preferred
Tigecycline is a broad-spectrum glycylcycline antibiotic, identified for use as a laboratory chemical and in the manufacture of substances. It exhibits inhibitory effects against various bacteria including *E. coli* and *Acinetobacter baumannii*, and has shown antileukemic activity in both in vitro and in vivo studies.
  • Broad-spectrum glycylcycline antibiotic.
  • Active against *E. coli* with a mean inhibitory concentration of 125 ng/mL.
  • Effective against *Acinetobacter baumannii* with MIC50 and MIC90 of 1 and 2 mg/L respectively.
  • Demonstrates inhibition of AML2 and HL-60 human leukemia cells.
  • Reduces tumor volume and weight in an AML xenograft model.
Catalog No. 50-004-50872