Learn More
Medchemexpress LLC TIGIT Mouse HEK293 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP77235500UG
Encompass_Preferred
TIGIT protein with signaling receptor binding activity functions upstream of T cell activation suppression Active on the cell surface it shares orthology with human TIGIT Associated with modulating T cell responses TIGIT exhibits biased expression notably in the large intestine small intestine and other tissues This highlights its potential in immune regulation within these contexts TIGIT Protein Mouse (HEK293 mFc) is the recombinant mouse-derived TIGIT protein expressed by HEK293 with C-mFc labeled tag
Catalog No. 50-001-86536