Learn More
Medchemexpress LLC Tigloylgomisin P | 69176-51-8 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN75861MG
Encompass_Preferred
Tigloylgomisin P is a lignin known for its anti-HIV activity, exhibiting an EC₅₀ of 37 μM. It also demonstrates an anticancer effect.
  • Acts as a lignin
  • Exhibits anti-HIV activity with an EC₅₀ of 37 μM in H9 cells
  • Demonstrates an anticancer effect
  • Shows weak cytotoxicity against A549 cells (GI₅₀=18.77 μM)
  • Shows weak cytotoxicity against epidermoid carcinoma of the nasopharynx (KB; GI₅₀=13.91 μM)
Catalog No. 50-004-17381