Learn More
Medchemexpress LLC Tiglyl carnitine (Standard) | 64681-36-3 | 98.01% | 243.30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113408R
Encompass_Preferred
Tiglyl carnitine (Standard) is an analytical standard intended for research and analytical applications. It is associated with celiac disease and mitochondrial acetoacetyl-CoA thiolase (T2) deficiency.
  • Analytical standard for Tiglyl carnitine
  • Suitable for research and analytical applications
  • Associated with celiac disease
  • Associated with mitochondrial acetoacetyl-CoA thiolase (T2) deficiency
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS techniques
Catalog No. 50-004-39065