Learn More
Cayman Chemical TIgR17trIfluoROCTAte sal 10mg
SDP
Supplier:  Cayman Chemical 3747310 MG
Encompass_Preferred
A cyclic peptide; increases proliferation of HaCaT keratinocytes and human skin fibroblasts at 5-20 UG/ml; increases HaCaT cell migration at 20 UG/ml; increases the rate of wound closure and re-epithelialization in a mouse model of full-thickness dermal wounds at 20 UG/ml, topically
Catalog No. 50-291-8092