Learn More
Cayman Chemical TIlmIcosInphosphate 10mg
SDP
Supplier:  Cayman Chemical 1819010 MG
Encompass_Preferred
A macrolide antibiotic; active against macrolide-susceptible strains of M. haemolytica, P. multocida, and E. coli (MICs = 4, 4, and 128 µM, respectively), as well as a hyperpermeable strain of E. coli (MIC = 2 µM); inhibits bacterial protein synthesis (IC50 = 0.36 µM); increases serum CK, CK-MB, and MDA levels in mice, indicating cardiotoxicity
Catalog No. 50-296-6742