Learn More
Selleck Chemical LLC Tilorone dihydrochloride S5020-25mg
SDP
Supplier:  Selleck Chemical LLC S502025MG
Encompass_Preferred
Tilorone dihydrochloride is a broad-spectrum orally active antiviral agent that activates interferon production It has antitumor anti-inflammatory properties
Catalog No. 50-311-2361