Learn More
Waters Corp Tilted Vial Domino, 10° Angle, 12-Vial Capacity
Supplier:  Waters Corp 186010572
Encompass_Preferred
The Tilted Vial Domino holds vials at a 10° angle to minimize dead volume during automated use, accommodating up to 12 vials. This machine-readable consumable holder interfaces with the Andrew+ pipetting robot for seamless laboratory automation. It features QR codes for labware information and ARUCO codes for spatial positioning.
  • Holds vials at 10° angle to minimize dead volume
  • Accommodates up to 12 vials
  • Machine-readable for automated workflows
  • Equipped with QR and ARUCO codes for positioning and information
  • Compatible with pipetting robot for seamless automation
Catalog No. NC3876160