Learn More
Medchemexpress LLC TIM-3/HAVCR2 Cynomo 50ug
SDP
Supplier:  Medchemexpress LLC HYP7244750UG
Encompass_Preferred
The TIM-3/HAVCR2 protein is a cell surface receptor that regulates immune responses by inhibiting macrophage activation and suppressing Th1-mediated autoimmunity TIM-3/HAVCR2 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived TIM-3/HAVCR2 protein expressed by HEK293 with N-8 His labeled tag
Catalog No. 50-001-81439