Learn More
Medchemexpress LLC TIM-4/TIMD-4 Mouse 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77236100UG
Encompass_Preferred
The TIM-4/TIMD-4 protein is a phosphatidylserine receptor that regulates immune responses through multiple actions It participates in phagocytosis of apoptotic cells and regulates T cell function TIM-4/TIMD-4 Protein Mouse (HEK293 His) is the recombinant mouse-derived TIM-4/TIMD-4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86565