Learn More
Research Products International Corp Time Tape, 3"" Core, 3/4"" Wide, Tan, 6/cs
SDP
Supplier:  Research Products International Corp 512605T
Encompass
Labeling tapes suited for a wide variety of laboratory uses. Easy to write on with pencil, pen, or permanent marker. Strong, durable tapes are resistant to oil, water, and acids. Withstand extreme temperature ranges from -10°F up to +250°F. Can be applied at temperatures as low as +32°F. Features a removable adhesive that peels off glass, plastic or metal.
Catalog No. 50-197-7904