Learn More
Research Products International Corp Luria Brth Millr Pwd Anml Free
Supplier: Research Products International Corp L24340100.0
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade. Dissolve 20 grams in 1 liter distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes.