Learn More
Strem, An Ascensus Company CAS# 7772-99-8. 100g. Tin(II) chloride, anhydrous, 98%. MFCD00011241
SDP
Supplier:  Strem, An Ascensus Company 935030100G
Encompass
CAS# 7772-99-8. 100g. Tin(II) chloride, anhydrous, 98%. MFCD00011241. Molecular Weight: 189.60. Molecular Formula: SnCl2. Color/form: white xtl. Strem# 93-5030. http://www.strem.com/catalog/v/93-5030/
Catalog No. 50-901-15582