Learn More
Strem, An Ascensus Company CAS# 10025-69-1. 500g. Tin(II) chloride dihydrate, 98%. MFCD00149863
SDP
Supplier:  Strem, An Ascensus Company 935031500G
Encompass
CAS# 10025-69-1. 500g. Tin(II) chloride dihydrate, 98%. MFCD00149863. Molecular Weight: 189.60 (225.63). Molecular Formula: SnCl2∙2H2O. Color/form: white xtl. Strem# 93-5031. http://www.strem.com/catalog/v/93-5031/
Catalog No. 50-901-15585