Learn More
Strem, An Ascensus Company CAS# 7646-78-8. 250g. Tin(IV) chloride, anhydrous, 98%. MFCD00011242
SDP
Supplier:  Strem, An Ascensus Company 935022250G
Encompass
CAS# 7646-78-8. 250g. Tin(IV) chloride, anhydrous, 98%. MFCD00011242. Molecular Weight: 260.50. Molecular Formula: SnCl4. Color/form: colorless liq. Strem# 93-5022. http://www.strem.com/catalog/v/93-5022/
Catalog No. 50-901-15570