Learn More
Strem, An Ascensus Company CAS# 36809-75-3. 5g. Tin(IV) t-butoxide (99.99%-Sn) PURATREM. MFCD00156551
SDP
Supplier:  Strem, An Ascensus Company 5021005G
Encompass
CAS# 36809-75-3. 5g. Tin(IV) t-butoxide (99.99%-Sn) PURATREM. MFCD00156551. Molecular Weight: 411.17. Molecular Formula: Sn(OC4H9)4. Color/form: white solid to turbid colorless liq. Strem# 50-2100. http://www.strem.com/catalog/v/50-2100/
Catalog No. 50-901-12616