Learn More
Cayman Chemical TIn MSoporphyrIn IXCLOrId 10mg
SDP
Supplier:  Cayman Chemical 1907110 MG
Encompass_Preferred
A potent, competitive inhibitor of HO activity in vitro (Ki = 14 nM); inhibits hepatic, renal, and splenic HO activity in vivo for extended periods of time; blocks bilirubin production in vivo, decreasing HO activity in animal models of hyperbilirubinemia
Catalog No. 50-298-2584