Learn More
Medchemexpress LLC HY-101194 5mg Medchemexpress, Tin-protoporphyrin IX CAS:14325-05-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011945MG
Encompass_Preferred
Medchemexpress, HY-101194 5mg Tin-protoporphyrin IX CAS:14325-05-4 Tin-protoporphyrin IX (SnPPIX) is a potent Heme oxygenase-1 (HO-1) inhibitor. Tin-protoporphyrin IX (SnPPIX) sensitizes pancreatic ductal adenocarcinoma (PDAC) tumors to chemotherapy in mice model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0620