Learn More
Medchemexpress LLC Tin protoporphyrin IX dichloride | 14325-05-4 | 96.2% | 750.26 g/mol | C34H32Cl2N4O4Sn | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101194100MG
Encompass_Preferred
Tin protoporphyrin IX dichloride is a metalloporphyrin research reagent that acts as a potent inhibitor of heme oxygenase enzymes. It is provided as a milligram-scale crystalline solid intended for biochemical and cell-based studies to probe heme metabolism, oxidative stress, and related pathways.
  • Potent inhibitor of heme oxygenase enzymes.
  • Suitable for in vitro and in vivo research applications.
  • Available in milligram-scale sizes, including 100 MG.
  • High reported purity for research use (approximately 96.2%).
  • Supplied as a stable solid for routine laboratory handling.
Catalog No. 50-002-12075