Learn More
Medchemexpress LLC Tin protoporphyrin IX dichloride | 14325-05-4 | 96.2% | 750.26 g/mol | C34H32Cl2N4O4Sn | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10119425MG
Encompass_Preferred
Tin protoporphyrin IX dichloride is a tin-containing porphyrin analog used as a research reagent to inhibit heme oxygenase-1 (HO-1) and study heme metabolism and related cellular pathways.
  • Potent heme oxygenase-1 (HO-1) inhibitor for mechanistic studies.
  • Suitable for biochemical and cellular assays.
  • High purity (96.2%) for reproducible results.
  • Chemical formula C34H32Cl2N4O4Sn; molecular weight 750.26 g/mol.
  • Available in multiple vial sizes, including 25 mg.
  • Store sealed, away from moisture; in solvent, -80°C for up to 6 months.
Catalog No. 50-002-12074